Kupfer(I)bromid-Dimethylsulfid-Komplex 99%
Lieferant: Thermo Fisher Scientific
213180100
213180250
213181000
ACRO213180100EA
91.2
EUR
ACRO213180100
ACRO213180250
ACRO213181000
Kupfer(I)bromid-Dimethylsulfid-Komplex 99%
Kupfer(I)bromid-Dimethylsulfid-Komplex
Formel:
C₂H₆BrCuS Schmelzpunkt: 132 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043295 CAS-Nummer: 54678-23-8 EINECS: 259-292-5 |
Spezifikation Testergebnisse
Appearance | Off-white to beige to green to grey Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Titration Iodometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...