Methyl-1-cyclohexene-1-carboxylate 98%

Lieferant: Thermo Scientific

Synonyme: Methyl cyclohex-1-enecarboxylate

213710050
ACRO213710050EA 109 EUR
ACRO213710050
Methyl-1-cyclohexene-1-carboxylate 98%
Methyl-1-cyclohexene-1-carboxylate

Appearance: Clear colorless Liquid

Formel: C₈H₁₂O₂
Molecular Weight: 140,182 g/mol
Siedepunkt: 190…192 °C (760 mmHg)
Dichte: 1,03 g/cm³
Flammpunkt: 73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001544
CAS-Nummer: 18448-47-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR