Molekularsieb A5 (0,5 nm, 5 Å), Pulver < 50 µm

Lieferant: Thermo Scientific

Synonyme: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets

214815000
ACRO214815000EA 63.3 EUR
ACRO214815000
Molekularsieb A5 (0,5 nm, 5 Å), Pulver < 50 µm
Molekularsieb A5 (0,5 nm, 5 Å)
MDL: MFCD00131613
CAS-Nummer: 69912-79-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
pH 10 to 10.8 (10 % suspension in water)
Residual water ≤1.5 % (550°C, 2 h)
Water adsorption ≥23 % (50%rH, 20°C, 24h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR