Serotonin Hydrochlorid 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
215021000
215025000
ACRO215021000EA
142
EUR
ACRO215021000
ACRO215025000
Serotonin Hydrochlorid 97+%
Serotonin Hydrochlorid
Formel:
C₁₀H₁₃ClN₂O Schmelzpunkt: 168…172 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00012686 CAS-Nummer: 153-98-0 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Loss on drying | ≤2.5 % (115°C, 3 hrs) (vacuum) |
Appearance of solution | colorless to light yellow |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...