Eriochromcyanin R, rein

Lieferant: Thermo Scientific

Synonyme: Chromoxane Cyanine R

215150050
ACRO215150050EA 21.8 EUR
ACRO215150050
Eriochromcyanin R, rein
Eriochromcyanin R
Formel: C₂₃H₁₅Na₃O₉S
Lagertemperatur: Raumtemperatur
MDL: MFCD00012411
CAS-Nummer: 3564-18-9
EINECS: 222-641-7

Bestellen

Spezifikation Testergebnisse

Appearance Red to dark brown Powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR