Iron(III) hexacyanoferrate(II), rein

Lieferant: Thermo Fisher Scientific

215210250 215211000
ACRO215210250EA 53.8 EUR
ACRO215210250 ACRO215211000
Iron(III) hexacyanoferrate(II), rein
Iron(III) hexacyanoferrate(II)

CAS No.: 14038-43-8

Formel: C₁₈Fe₇N₁₈
Lagertemperatur: Raumtemperatur
MDL: MFCD00135663
CAS-Nummer: 14038-43-8

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR