Rhodamine B base 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Xanthylium, 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-, inner salt, 9-(2-Carboxylatophenyl)-3,6-bis(diethylamino)xanthylium, Ethaminium, N-(9-(2-carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene)-N-ethyl)-, hydroxide, inner salt
215230250
215231000
ACRO215230250EA
79.9
EUR
ACRO215230250
ACRO215231000
Rhodamine B base 97%
Rhodamine B base
CAS No.: 509-34-2
Formel:
C₂₈H₃₀N₂O₃ Molecular Weight: 442.559 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066967 CAS-Nummer: 509-34-2 UN: 3077 |
Spezifikation Testergebnisse
Appearance | Pink to red to violet Powder and/or chunks |
Infrared spectrum | Conforms |
Dye content | ≥96.0 % |
UV Lambda max | 542 to 546 nm (in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...