Natriumhydrogencarbonat 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific

217120010 217120025 217125000 217120100
ACRO217120010EA 43.8 EUR
ACRO217120010 ACRO217120025 ACRO217125000 ACRO217120100
Natriumhydrogencarbonat 99.5% zur Analyse
Natriumhydrogencarbonat
Formel: NaHCO₃
Schmelzpunkt: 270 °C
Dichte: 2,159 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003528
CAS-Nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Titration with HCl ≥99.4 %
Chloride (Cl) ≤100 ppm
Phosphate (PO4) ≤30 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤2 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Zinc (Zn) ≤5 ppm
Insoluble matter ≤0.015 %
Total sulfur (S) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR