(S)-(-)-1-(1-Naphthyl)ethylamin 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(-)-alpha-(1-Naphthyl)ethylamine

217810010
ACRO217810010EA 36.5 EUR
ACRO217810010
(S)-(-)-1-(1-Naphthyl)ethylamin 99+%
(S)-(-)-1-(1-Naphthyl)ethylamin
Formel: C₁₂H₁₃N
Molecular Weight: 171,24 g/mol
Siedepunkt: 153 °C (11 mmHg)
Dichte: 1,06 g/cm³
Flammpunkt: >112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064179
CAS-Nummer: 10420-89-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6215 to 1.6245 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation -51° to -59° (20°C, 589 nm) (c=2, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR