L(+)-α-Phenylglycin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Phg-OH, L-(+)-2-Phenylglycine

217990250 217991000
ACRO217990250EA 45 EUR
ACRO217990250 ACRO217991000
L(+)-α-Phenylglycin 99%
L(+)-α-Phenylglycin
Formel: C₈H₉NO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064403
CAS-Nummer: 2935-35-5
EINECS: 220-909-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 h)
Specific optical rotation +154° to +160° (20°C, 589 nm) (c=1, 1 M HCl)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR