D-(-)-Pantolactone 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pantolactone, (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, D-(-)-Pantoyl Lactone, D-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone
218140250
218140050
218141000
ACRO218140250EA
70.9
EUR
ACRO218140250
ACRO218140050
ACRO218141000
D-(-)-Pantolactone 99%
D-(-)-Pantolactone
CAS No.: 599-04-2
Formel:
C₆H₁₀O₃ Siedepunkt: 120…122 °C (15 mmHg) Schmelzpunkt: 88…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005392 CAS-Nummer: 599-04-2 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 88 to 92 °C |
GC | ≥98.5 % |
Specific optical rotation | -49° to -52° (25 °C, 589 nm) (c=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...