Ammoniumacetat 98+% zur Analyse

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Essigsäure Ammoniumsalz

218360010 218365000 218360050
ACRO218360010EA 86.3 EUR
ACRO218360010 ACRO218365000 ACRO218360050
Ammoniumacetat 98+% zur Analyse
Ammoniumacetat
Formel: H₃CCOONH₄
Schmelzpunkt: 114 °C
Dichte: 1,17 g/cm³ (20 °C)
Flammpunkt: 136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013066
CAS-Nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤3.0 %
Ash ≤0.01 %
pH 6.5 to 7.5 (5 % at 20°C)
Titration excess NaOH ≥98.0 % (on dry substance)
Chloride (Cl) ≤20 ppm
Nitrate (NO3) ≤20 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR