Calciumsulfat, Drierite™ 4 mesh for drying, ohne Indikator
Lieferant: Thermo Fisher Scientific
Synonyme:
Calcium sulphate (anhydrous) anhydrite, Gypsum
219030020
219035000
219030100
ACRO219030020EA
204
EUR
ACRO219030020
ACRO219035000
ACRO219030100
Calciumsulfat, Drierite™ 4 mesh for drying, ohne Indikator
Calcium sulphate
Formel:
CaO₄S Schmelzpunkt: 1450 °C Dichte: 2,964 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010912 CAS-Nummer: 7778-18-9 EINECS: 231-900-3 Merck Index: 13,01711 |
Spezifikation Testergebnisse
Appearance | White to greyish-white Irregular granules |
Water | ≤0.3 % |
Particle size | 4 mesh |
Composition | ≥99.5 % (CaSO4) Typical |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...