Thioharnstoff 99+% zur Analyse
Lieferant: Thermo Fisher Scientific
|
Achtung
|
220050010
220052500
220050050
ACRO220050010EA
83.9
EUR
ACRO220050010
ACRO220052500
ACRO220050050
Thioharnstoff 99+% zur Analyse
Thioharnstoff
Formel:
NH₂CSNH₂ Schmelzpunkt: 171…175 °C Dichte: 1,435 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008067 CAS-Nummer: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 13,09443 |
|
Spezifikation Testergebnisse
Appearance | White Crystals |
Infrared spectrum | Conforms |
Titration Iodimetric | ≥99.0 % |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
Sulfate (SO4) | ≤50 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤10 ppm |
Zinc (Zn) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...