Kupfer(II)oxid 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
221281000
221280025
221285000
ACRO221281000EA
49
EUR
ACRO221281000
ACRO221280025
ACRO221285000
Kupfer(II)oxid 99+%
Kupfer(II)oxid
Formel:
CuO Siedepunkt: 2000 °C (1013 hPa) Schmelzpunkt: 1326 °C Dichte: 4,9 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010979 CAS-Nummer: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9,III Merck Index: 13,02674 |
Spezifikation Testergebnisse
Appearance | Black Powder |
Titration Complexometric | ≥99.0 % |
Chloride (Cl) | ≤50 ppm |
Carbon (C) | ≤500 ppm |
Iron (Fe) | ≤500 ppm |
Total sulfur (as SO4) | ≤0.02 % |
Free alkali | Passes test |
Insoluble matter | ≤0.02 % (in dilute HCl) |
Nitrogen compounds (as N) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...