(Brommethyl)cyclopropan 96%

Lieferant: Thermo Fisher Scientific
Achtung

221810010 221810050 221810250
ACRO221810010EA 81.4 EUR
ACRO221810010 ACRO221810050 ACRO221810250
(Brommethyl)cyclopropan 96%
(Brommethyl)cyclopropan
Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 101…108 °C (760 mmHg)
Dichte: 1.38 g/cm³
Flammpunkt: 41 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥95 %
Specific gravity (20°C) 1.385 to 1.415


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR