Tetrakis(triphenylphosphin)platin(0) 98%

Lieferant: Thermo Fisher Scientific

223960010 223960050
ACRO223960010EA 124 EUR
ACRO223960010 ACRO223960050
Tetrakis(triphenylphosphin)platin(0) 98%
Tetrakis(triphenylphosphin)platin(0)
Formel: C₇₂H₆₀P₄Pt
Schmelzpunkt: 159…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00010014
CAS-Nummer: 14221-02-4
EINECS: 238-087-4

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Carbon (C) 67.8 to 71.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR