(3,4-Dihydroxyphenyl)essigsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

225600010 225600050 225600250
ACRO225600010EA 38.8 EUR
ACRO225600010 ACRO225600050 ACRO225600250
(3,4-Dihydroxyphenyl)essigsäure 98%
(3,4-Dihydroxyphenyl)essigsäure
Formel: (C₆H₃(OH)₂)CH₂COOH
Molecular Weight: 168.15 g/mol
Siedepunkt: 337
Schmelzpunkt: 126…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004338
CAS-Nummer: 102-32-9
EINECS: 203-024-1

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light brown to purple-grey Powder
Infrared spectrum Conforms
Melting point 127 to 132 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR