Palatinose hydrate 98%

Lieferant: Thermo Fisher Scientific

225950250 225951000 225955000
ACRO225950250EA 36.2 EUR
ACRO225950250 ACRO225951000 ACRO225955000
Palatinose hydrate 98%
Palatinose hydrate
Formel: C₁₂H₂₂O₁₁·xH₂O
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076094
CAS-Nummer: 343336-76-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤6 %
HPLC ≥97.5 % (ELSD)
Specific optical rotation +97° to +101° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR