Guanosin-5'-triphosphat Dinatriumsalz Hydrat 90% für die Biochemie
Lieferant: Thermo Fisher Scientific
226250010
226250500
226252500
ACRO226250010EA
566
EUR
ACRO226250010
ACRO226250500
ACRO226252500
Guanosin-5'-triphosphat Dinatriumsalz Hydrat 90% für die Biochemie
Guanosin-5'-triphosphat Dinatriumsalz Hydrat
Formel:
C₁₀H₁₄N₅Na₂O₁₄P₃·H₂O Lagertemperatur: Gefrierschrank |
MDL:
MFCD03410297 CAS-Nummer: 56001-37-7 EINECS: 259-940-7 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤12 % (K.F.) |
HPLC | ≥88 % |
UV spectrophotometry | ≥88 % (lambda max. 252 nm, pH 7) |
Ratio of Absorbances | 0.63 to 0.69 (A280/A260) |
Related substances | ≤4 % (typically) Guanosine diphosphate (HPLC) |
Sodium (Na) | 7 to 9.5 % (flame photom.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...