Inosine 5'-monophosphate disodium salt hydrate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

226260050 226260250 226261000
ACRO226260050EA 40.9 EUR
ACRO226260050 ACRO226260250 ACRO226261000
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate

CAS No.: 352195-40-5

Formel: C₁₀H₁₁N₄Na₂O₈P·xH₂O
Molecular Weight: 392.17 g/mol (anhy)
Schmelzpunkt: 175 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00150372
CAS-Nummer: 352195-40-5

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤28.5 % (K.F.)
UV spectrophotometry molar absorptivity 12300 +/- 4%, on dry basis
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR