Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) 90%

Lieferant: Thermo Fisher Scientific

Synonyme: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

226310010
ACRO226310010EA 202 EUR
ACRO226310010
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) 90%
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Formel: C₉H₁₂N₂Na₃O₁₅P₃
Molecular Weight: 550.09 g/mol
Schmelzpunkt: >140 °C (Zers.)
Flammpunkt: >110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00150714
CAS-Nummer: 19817-92-6
EINECS: 243-347-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤15 % (105°C, 5 hrs)
HPLC ≥88 %
UV Lambda max 262 nm (+/- 2 nm)
pH 3.0 to 7.0 (0.5 % in water)
Absorbance scale 0.70 to 0.80 (A250/A260)
Chloride (Cl) ≤2000 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR