6-Benzylaminopurin 99%

Lieferant: Thermo Fisher Scientific
Achtung

226410010 226410050 226410250
ACRO226410010EA 36.4 EUR
ACRO226410010 ACRO226410050 ACRO226410250
6-Benzylaminopurin 99%
6-Benzylaminopurin
Formel: C₁₂H₁₁N₅
Schmelzpunkt: 229…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005572
CAS-Nummer: 1214-39-7
EINECS: 214-927-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 229 to 233 °C
Loss on drying ≤0.5 %
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR