6-Chlor-9-(β-D-ribofuranosyl)-9H-purin 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Chloropurine riboside

226450010 226450050
ACRO226450010EA 119 EUR
ACRO226450010 ACRO226450050
6-Chlor-9-(β-D-ribofuranosyl)-9H-purin 99+%
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

CAS No.: 5399-87-1

Formel: C₁₀H₁₁ClN₄O₄
Molecular Weight: 286.67 g/mol
Schmelzpunkt: 161…163 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00005738
CAS-Nummer: 5399-87-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR