6-Mercaptopurine monohydrate 99.5+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate

226520050 226520250
ACRO226520050EA 81.2 EUR
ACRO226520050 ACRO226520250
6-Mercaptopurine monohydrate 99.5+%
6-Mercaptopurine monohydrate

Appearance: Yellow Powder

Formel: C₅H₄N₄S
Schmelzpunkt: 313 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01461928
CAS-Nummer: 6112-76-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Water 9 to 13 % (K.F.)
HPLC ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR