6-(Methylthio)purin 98%
Lieferant: Thermo Fisher Scientific
226560010
ACRO226560010EA
79
EUR
ACRO226560010
6-(Methylthio)purin 98%
6-(Methylthio)purine
Formel:
C₆H₆N₄S Schmelzpunkt: 219…223 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005576 CAS-Nummer: 50-66-8 EINECS: 200-057-3 |
Spezifikation Testergebnisse
Appearance | White to light yellow to green-beige Powder |
Infrared spectrum | Conforms |
Melting point | 219 to 223 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...