Thymidin (2'-Deoxythymidin) 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

226740050 226740250 226741000
ACRO226740050EA 165 EUR
ACRO226740050 ACRO226740250 ACRO226741000
Thymidin (2'-Deoxythymidin) 99+%
Thymidin (2'-Deoxythymidin)
Formel: C₁₀H₁₄N₂O₅
Molecular Weight: 242.23 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006537
CAS-Nummer: 50-89-5
EINECS: 200-070-4

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.3 % (800°C)
HPLC ≥99.0 %
UV spectrophotometry molar absorptivity 9600 +/- 4%, on dry basis
Ratio of Absorbances 0.70 to 0.76 (A280/A260)
Specific optical rotation +17.5° to +19.5° (20°C, 589 nm) (c=3, H2O)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR