5-Amino-2,4(1H,3H)-pyrimidindion 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Aminouracil, 5-Amino-2,4-dihydroxypyrimidin
226820250
ACRO226820250EA
111
EUR
ACRO226820250
5-Amino-2,4(1H,3H)-pyrimidindion 98%
5-Aminouracil
Formel:
C₄H₅N₃O₂ Molecular Weight: 127,1 g/mol Siedepunkt: 377 °C (1013 hPa) Schmelzpunkt: >300 °C |
MDL:
MFCD00006025 CAS-Nummer: 932-52-5 EINECS: 213-252-3 |
Spezifikation Testergebnisse
Appearance | Beige to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...