N-Acetyl-DL-Tryptophan 98+%, reinst
Lieferant: Thermo Fisher Scientific
Synonyme:
Ac-DL-Trp-OH, N(α)-Acetyl-DL-Tryptophan
227150250
227151000
ACRO227150250EA
29.1
EUR
ACRO227150250
ACRO227151000
N-Acetyl-DL-Tryptophan 98+%, reinst
N-Acetyl-DL-Tryptophan
Formel:
C₁₃H₁₄N₂O₃ Molecular Weight: 246.27 g/mol Schmelzpunkt: 204…206 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005644 CAS-Nummer: 87-32-1 EINECS: 201-739-3 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C) |
Sulfated ash | ≤0.2 % |
Titration excess NaOH | 98.0 to 101.0 % (On dry substance) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...