Cytosine-β-D-arabinofuranoside hydrochloride 99%, kristallin

Lieferant: Thermo Scientific
Achtung

Synonyme: Arabinosylcytosine hydrochloride, Cytarabine HCI, Cytarabine HCl, 4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride, 1-(β-D-Arabinofuranosyl)cytosine HCl

227180010
ACRO227180010EA 183 EUR
ACRO227180010
Cytosine-β-D-arabinofuranoside hydrochloride 99%, kristallin
Cytosine-beta-D-arabinofuranoside hydrochloride
Formel: C₉H₁₄ClN₃O₅
Molecular Weight: 279,677 g/mol
Schmelzpunkt: 197…198 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012839
CAS-Nummer: 69-74-9

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +127° to +133° (20°C, 589 nm) (c=0.7, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR