β-Cyclodextrin Hydrat 99%

Lieferant: Thermo Fisher Scientific

227280050 227280250 227281000
ACRO227280050EA 27.1 EUR
ACRO227280050 ACRO227280250 ACRO227281000
β-Cyclodextrin Hydrat 99%
β-Cyclodextrin Hydrat
Formel: C₄₂H₇₀O₃₅·H₂O
Schmelzpunkt: 298…300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150811
CAS-Nummer: 68168-23-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤14 % (K.F.)
Heavy metals ≤10 ppm
Sulfated ash ≤0.1 %
Solubility (1 % in water) Clear colorless
HPLC ≥98.5 % (ELSD)
Related substances ≤1.0 % Alpha- and gamma-cyclodextrin (HPLC)
pH 5.0 to 8.0 (1 % in water)
Specific optical rotation +155° to +167° (25°C, 589 nm) (c=1, H20) (On dry substance)
Reducing substances ≤0.2 % (as Glucose)
Residual solvents ≤5 ppm Toluene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR