Aflatoxin B1, kristallin

Lieferant: Thermo Fisher Scientific
Gefahr

227340010 227340100 227340500
ACRO227340010EA 58.5 EUR
ACRO227340010 ACRO227340100 ACRO227340500
Aflatoxin B1, kristallin
Aflatoxin B1
Formel: C₁₇H₁₂O₆
Molecular Weight: 312.3 g/mol
Schmelzpunkt: 268…269 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00869647
CAS-Nummer: 1162-65-8
UN: 3462
ADR: 6.1 I

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Solubility (10 mg/ml in CH2Cl2) Clear colorless to light yellow
HPLC ≥98 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR