Auramin O, rein biologische Färbemittel
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
227520500
ACRO227520500EA
64.5
EUR
ACRO227520500
Auramin O, rein biologische Färbemittel
Auramin O
Formel:
C₁₇H₂₂ClN₃ Molecular Weight: 303,84 g/mol Schmelzpunkt: >250 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012484 CAS-Nummer: 2465-27-2 EINECS: 219-567-2 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow to ochre Powder |
Infrared spectrum | Conforms |
Dye content | ≥60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...