Polyvinylpyrrolidon, M.W. K30 (MW ∼ 50.000)

Lieferant: Thermo Fisher Scientific

227541000 227545000 227540025
ACRO227541000EA 49.8 EUR
ACRO227541000 ACRO227545000 ACRO227540025
Polyvinylpyrrolidon, M.W. K30 (MW ∼ 50.000)
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Schmelzpunkt: 225 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellowish-white Hygroscopic powder
Infrared spectrum Conforms
Water ≤5.0 % (K.F.)
Sulfated ash ≤0.1 %
HPLC ≤3.0 % (2-Pyrrolidone)
Appearance of solution (5 % in water at 25°C) Clear
Viscosity 27 to 32 K-value (1 % w/v in water at 25°C)
pH 3.0 to 5.0 (5 % in water)
Nitrogen (N) 11.5 to 12.8 % (on anhydrous substance)
Aldehyde (C2H4O) ≤0.05 %
Hydrazine (N2H4) ≤1 ppm
Peroxides ≤400 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR