trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%

Lieferant: Thermo Fisher Scientific
Achtung

228040500 228042500
ACRO228040500EA 158 EUR
ACRO228040500 ACRO228042500
trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₅NO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001466
CAS-Nummer: 1197-18-8
EINECS: 214-818-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 6.5 to 8.0 (5 % solution)
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR