4-Aminobenzamidin Dihydrochlorid 97%

Lieferant: Thermo Fisher Scientific
Achtung

228050050 228050010 228050250
ACRO228050050EA 92.1 EUR
ACRO228050050 ACRO228050010 ACRO228050250
4-Aminobenzamidin Dihydrochlorid 97%
4-Aminobenzamidin Dihydrochlorid
Formel: C₇H₁₁Cl₂N₃
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013001
CAS-Nummer: 2498-50-2
EINECS: 219-692-2

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR