m-Kresolpurpur, rein Indikator-Qualität

Lieferant: Thermo Fisher Scientific

228200050 228200100 228200500
ACRO228200050EA 45.4 EUR
ACRO228200050 ACRO228200100 ACRO228200500
m-Kresolpurpur, rein Indikator-Qualität
m-Kresolpurpur
Formel: C₂₁H₁₈O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00005871
CAS-Nummer: 2303-01-7
EINECS: 218-960-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Solubility (0.1 % in 50 % ethanol) Clear
Specific extinction E (1%/1cm) ≥900 (578 nm, in buffer)
Visual transition interval from pH 7.4 (yellow) to pH 9.0 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR