4(3H)-Pyrimidinone 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Pyrimidinol, 4(3H)-Pyrimidone, 3H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(3H)-one

228250100
ACRO228250100EA 141 EUR
ACRO228250100
4(3H)-Pyrimidinone 98+%
4(3H)-Pyrimidinone

Appearance: White to light yellow Powder

Formel: C₄H₄N₂O
Molecular Weight: 96,09 g/mol
Schmelzpunkt: 163…168 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006664
CAS-Nummer: 51953-17-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR