D-Cycloserin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: D(+)-Cycloserin, D-4-Aminoisoxazolidin-3-on, (R)-(+)-Cycloserin

228480010 228480050 228480250
ACRO228480010EA 73 EUR
ACRO228480010 ACRO228480050 ACRO228480250
D-Cycloserin 98%
D-Cycloserin
Formel: C₃H₆N₂O₂
Molecular Weight: 102.09 g/mol
Schmelzpunkt: 147 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00005353
CAS-Nummer: 68-41-7
EINECS: 200-688-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 % (60°C, 3 hrs) (vacuum)
UV at 285 nm A: ≤0.8
Condensation products (0.04 % in 0.1 N NaOH)
Potency ≥900 µg/mg
Residue after ignition ≤0.5 %
pH 5.5 to 6.5 (10 % solution)
Specific optical rotation +108° to +114° (20°C, 589 nm) (c=5, 2N NaOH)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR