5-Brom-2'-deoxyuridin (Broxuridin) 99+%

Lieferant: Thermo Fisher Scientific
Achtung

228590010 228590025 228595000 228590100
ACRO228590010EA 87.4 EUR
ACRO228590010 ACRO228590025 ACRO228595000 ACRO228590100
5-Brom-2'-deoxyuridin (Broxuridin) 99+%
5-Brom-2'-deoxyuridin (Broxuridin)
Formel: C₉H₁₁BrN₂O₅
Schmelzpunkt: 191…194 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥99.0 %
Ratio of Absorbances 1.45 to 1.55 (A290/A260)
Microbial contamination ≤100 CFU/g (Total count)
Bacterial endotoxins ≤0.1 EU/mg (LAL-test)
Related substances ≤1.0 % (HPLC) Total impurities
Specific optical rotation +20° to +24° (20°C, 589 nm) (c=1, H2O)
Heavy metals (as Pb) ≤10 ppm
Residual solvents ≤0.1 % (GC) Methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR