D(+)-Prolin 99+%

Lieferant: Thermo Fisher Scientific

228660010 228660050 228660250
ACRO228660010EA 41.2 EUR
ACRO228660010 ACRO228660050 ACRO228660250
D(+)-Prolin 99+%
D(+)-Prolin
Formel: C₅H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +83.0° to +87.0° (20°C, 589 nm) (c=4, water)
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR