Creatinin 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
228940250
228940500
228942500
ACRO228940250EA
61.3
EUR
ACRO228940250
ACRO228940500
ACRO228942500
Creatinin 98%
Creatinin
Formel:
C₄H₇N₃O Molecular Weight: 113.12 g/mol Schmelzpunkt: 255 °C (Zers.) Flammpunkt: 290 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059730 CAS-Nummer: 60-27-5 EINECS: 200-466-7 |
Spezifikation Testergebnisse
Appearance | White to yellowish-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (105°C) |
Sulfated ash | ≤0.2 % |
Solubility | (5 % in water) Clear colorless to slightly yellow |
Titration with HClO4 | 97.5 to 102.5 % (on dried substance) |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...