L-Valine methyl ester hydrochloride 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl

229260100 229260500
ACRO229260100EA 53.8 EUR
ACRO229260100 ACRO229260500
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride

Appearance: White Powder or crystals

Formel: C₆H₁₄ClNO₂
Schmelzpunkt: 171…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012497
CAS-Nummer: 6306-52-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +14° to +17° (20°C, 589 nm) (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR