Kristallviolett, rein, hohe Reinheit biologische Färbemittel

Lieferant: Thermo Fisher Scientific
Gefahr

229641000 229645000
ACRO229641000EA 73.2 EUR
ACRO229641000 ACRO229645000
Kristallviolett, rein, hohe Reinheit biologische Färbemittel
Kristallviolett
Formel: C₂₅H₃₀ClN₃
Dichte: 1,19 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011750
CAS-Nummer: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to green-brown Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤9 % (110°C)
Sulfated ash ≤3 %
Titration ≥96.0 % (on dried substance)
Lead (Pb) ≤30 ppm
Zinc (Zn) Passes test
Insoluble matter (in ethanol) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR