Erioglaucine disodium salt, rein

Lieferant: Thermo Fisher Scientific

229730050 229730250
ACRO229730050EA 35 EUR
ACRO229730050 ACRO229730250
Erioglaucine disodium salt, rein
Erioglaucine disodium salt

Appearance: Dark purple Crystalline powder

Formel: C₃₇H₃₆N₂O₉S₃
Schmelzpunkt: 283 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012141
CAS-Nummer: 3844-45-9

Bestellen

Spezifikation Testergebnisse

Appearance Dark purple Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10 % (105°C)
UV Lambda max 625 to 630 nm
Absorption Ratio P-15/P+15 1.10 to 1.25
Specific absorbance A (1%/1cm) ≥1000 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR