Orange G 85+%, pure, high purity stain_biological

Lieferant: Thermo Fisher Scientific

Synonyme: Acid Orange 10, Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat, Orange G Natriumsalz, CI 16230

229820250 229821000
ACRO229820250EA 60.9 EUR
ACRO229820250 ACRO229821000
Orange G 85+%, pure, high purity stain_biological
Orange G
Formel: C₁₆H₁₀N₂Na₂O₇S₂
Molecular Weight: 452.38 g/mol
Schmelzpunkt: 141
Lagertemperatur: Raumtemperatur
MDL: MFCD00012457
CAS-Nummer: 1936-15-8
EINECS: 217-705-6

Bestellen

Spezifikation Testergebnisse

Appearance Orange to orange-red Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR