8-Azaguanine
Lieferant: Thermo Fisher Scientific
|
Achtung
|
229920010
229920050
ACRO229920010EA
88.4
EUR
ACRO229920010
ACRO229920050
8-Azaguanine
8-Azaguanine
- Bioactive
Formel:
C₄H₄N₆O Schmelzpunkt: >300 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00056937 CAS-Nummer: 134-58-7 |
Spezifikation Testergebnisse
Appearance | Yellow to light orange Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...