3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydrochloride TMB, TMB-HCl

229990010 229990050
ACRO229990010EA 64.2 EUR
ACRO229990010 ACRO229990050
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 98+%
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel: C₁₆H₂₀N₂·2HCl·xH₂O
Molecular Weight: 313.27 g/mol (anhy)
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.0 % (on dry substance)
Water ≤12.0 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR