3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydrochloride TMB, TMB-HCl
229990010
229990050
ACRO229990010EA
64.2
EUR
ACRO229990010
ACRO229990050
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 98+%
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel:
C₁₆H₂₀N₂·2HCl·xH₂O Molecular Weight: 313.27 g/mol (anhy) Schmelzpunkt: >300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00150104 CAS-Nummer: 207738-08-7 EINECS: 264-769-6 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Off-white Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.0 % (on dry substance) |
Water | ≤12.0 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...