Dexamethason 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum

230300010 230302500 230300050
ACRO230300010EA 126 EUR
ACRO230300010 ACRO230302500 ACRO230300050
Dexamethason 96%
Dexamethason
Formel: C₂₂H₂₉FO₅
Schmelzpunkt: 255…261 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064136
CAS-Nummer: 50-02-2
EINECS: 200-003-9

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (500 mg, 105°C)
UV spectrophotometry ≥95 % (On dried substance)
Related substances (HPLC) Passes test
Specific optical rotation +86° to +92° (20°C, 589 nm) (c=1, ethanol) (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR